0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
Furniture
Storage Cabinets
Accessories for storehouses for gas cylinders GLH
Accessories for storehouses for gas cylinders GLH
Accessories for storehouses for gas cylinders GLH
Catalog # LACOB80-5060-C
Supplier:
LACONT UMWELTTECHNIK
Expand image
Accessories for storehouses for gas cylinders GLH
Catalog # LACOB80-5060-C
Supplier:
LACONT UMWELTTECHNIK
Specifications
Description:
Ramp, drive-on, galvanised, with dimensions 1200×1000 mm
Seq:
01
Pk:
1
For:
GLH
Specifications
About this item
Product Details
Product Details