Order Entry
Hungary
Orders LinkContactUsLinkComponent
Colour test kit

Colour test kit

Colour test, model CO-1, 0-100 and 0-500, 1 KIT
Catalog # HACH223400
Supplier:   Hach
Colour test kit
Catalog # HACH223400
Supplier:  Hach
CAS Number:  
Colour test kit
Colour test kit

Colour test, model CO-1, 0-100 and 0-500, 1 KIT

CAS Number:  
Price
80 200 Ft
Log in for availability
1 * 1 KIT
Product Details & Documents

Colour disc kit reads apparent colour created by dissolved substances that can indicate industrial, agricultural or natural pollution. Dual range measurement in colour units.

Delivery information: Includes colour disc, comparator box and tubes, instructions and carrying case.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Colour test, model CO-1
  • Range:
    0-100 and 0-500

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.