Vinyl gloves, Rotiprotect®
Supplier: Roth Carl
Certificates
About this item
Flexible, ambidextrous gloves made of PVC. Ideal for food handling.
- Non sterile, powdered/powder-free, smooth finish
- Very good sense of touch
- With roll-type edge
- Free of latex proteins
Certifications: EN 455, Cat. I; AQL 1,5
Food contact approved. Not suitable for foodstuffs containing grease.
Food contact approved. Not suitable for foodstuffs containing grease.
Specifications
- Catalog No:
- ROTH6178.1
- ROTHX355.1
- ROTH6179.1
- ROTH1904.1
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powdered
- Powder-free
- Powdered
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455
- EN 455
- EN 455
- EN 455
- Food contact approved:
- +
- +
- +
- +