Order Entry
Hungary
ContactUsLinkComponent
Staining jars acc. to Hellendahl
Staining jars acc. to Hellendahl
Catalog # KAVAN23253111
Supplier:  Kavalier
Staining jars acc. to Hellendahl
Catalog # KAVAN23253111
Supplier:  Kavalier

Specifications

  • Description:
    Staining dishes acc. to Hellendahl
  • W×D×H:
    58,5×58,5×100 mm
  • Pk:
    1

Specifications

About this item

Soda-lime glass.

These staining trough hold up to 16 slides (76×26 mm).