Autoclave Bags
Supplier: DELTALAB
Certificates
About this item
Autoclave resistant bags, printed with the biohazard graphic symbol and instructions of use in five languages.
Caution: Do not close the bags hermetically when placing them in the autoclave at 121 °C.
Frequently Bought Together