SOLA™ SPE Plates, 96-Well
Supplier: Thermo Fisher Scientific
Certificates
About this item
SOLA™ fritless 96-well SPE plates provide greater reproducibility with cleaner, more consistent extracts. SOLA™ products provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
- Higher levels of reproducibility
- Higher levels of extract cleanliness
- Reduced solvent requirements
- Increased sensitivity
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together