Order Entry
Hungary
Orders LinkContactUsLinkComponent
D(+)-Fucose

D(+)-Fucose

Glass bottle for solids, 1 g
Catalog # MOLE31642524-1G
Marketsource
Supplier:   Molekula
D(+)-Fucose
Catalog # MOLE31642524-1G
Supplier:  Molekula
Marketsource
CAS Number:  3615-37-0
D(+)-Fucose

Glass bottle for solids, 1 g

CAS Number:  3615-37-0
Price
120 000 Ft
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  alpha-D-Fucose
  • CAS Number:3615-37-0
  • Formula:C₆H₁₂O₅
  • MDL Number:MFCD00135603
  • EINECS:222-792-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.