Order Entry
Hungary
ContactUsLinkComponent
4-Bromobenzylamine hydrochloride
4-Bromobenzylamine hydrochloride
Catalog # MOLEM53227275
Supplier:  Molekula
CAS Number:  
4-Bromobenzylamine hydrochloride
Catalog # MOLEM53227275
Supplier:  Molekula
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :222.52 g/mol
EINECS:247-503-3
CAS Number:26177-44-6
Formula:C₇H₉BrClN
MDL Number:MFCD00012860