Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc sulphate
Zinc sulphate 0.05 mol/l (0.05 N) in aqueous solution
Zinc sulphate 0.05 mol/l (0.05 N) in aqueous solution
POLK29253.1000
:
SPEKTRUM-3D
:
Zinc sulphate 0.05 mol/l (0.05 N) in aqueous solution
POLK29253.1000
:
SPEKTRUM-3D
:
Pack type:
Glass bottle
Pk:
1.000 mL
Seq:
2865
:
:
161.45 g/mol
:
231-793-3
:
GHS09
:
Ambient
:
7733-02-0
:
O₄SZn
:
MFCD00011302