Wash bottles, narrow or wide neck
Supplier: VITLAB
Certificates
About this item
Made of LDPE, translucent, screw cap and spray tube from PP.
- Good chemical resistance
- Good temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.
Frequently Bought Together