ContactUsLinkComponent
Downstream-Process1920-760.jpg
Alternative buffer management strategies to achieve downstream process improvements
There has been significant change across the biomanufacturing industry over the last couple of decades, as increased complexity of biologics and a growing focus on smaller patient populations have led to a greater need for cost control and process efficiency.
As productivity has improved as a result, so too have titers and other areas of process development. These advancements in science and technology offer exciting promise in drug development; however, they also create challenges for those still utilizing legacy processes and systems.
For example, while buffer preparation and management can be carried out in multiple ways, an established approach is to manufacture buffers in house with the use of water-for-Injection (WFI) to hydrate powders in stainless-steel or single-use tanks. Although this method is established and ideal for large volumes, it requires significant infrastructure, including the footprint needed for large tanks and for warehouse space for holding as well as weighing and dispensing areas for raw materials.
There are other approaches to this critical bioprocessing step, although each call on capabilities and expertise that may not exist at some companies and/or are difficult to obtain. Understanding not only the alternative buffer management strategies available but also what resources are needed to execute them can help ensure you have the tools necessary to remain competitive in today’s growing bioprocessing industry.
Read more
solutions-for-bioprocessing-260x190.jpg
Solutions for bioprocessing
From the small-scale bench to full commercial manufacturing, we enable our customers to reach the market with new treatments for patients, faster and safer. Learn more about our offering for biopharmaceutical manufacturing.
Learn more
Author
Pranav-Vengsarkar-100x100.jpg
Dr. Pranav Vengsarkar
Manager, Research & Development – Biopharma Production
Pranav Vengsarkar, Ph.D., is Manager, Process Development at Avantor, focused on product and process development for new cGMP products and excipients, and development and design of single-use raw material delivery systems. Pranav holds a Bachelor of Chemical Engineering degree from ICT, Mumbai and a Ph.D. in Chemical Engineering from Auburn University.