United States
Lien pour commanderContactUsLinkComponent
mabs-a-1920x760.jpg
Downstream Process Intensification with Novel Chromatography Resins for Enhanced Monoclonal Antibody Purification
Downstream Process Intesification with Novel Chromatography Resins for Enhanced Monoclonal Antibody Purification | Avantor
In this presentation, we will demonstrate an integrated mAbs downstream purification process to enable significant throughput improvement while removing of critical impurities.
The process approach includes (1) pre-absorption filtration technology and (2) buffer additives. Through use of pre-absorption filtration technology, in combination with PROchievA protein A affinity resin, that possess high dynamic binding capacities of greater than 60 g/L, we have obtained antibodies with increased purity, while reducing overall process time.
Furthermore, by using select additives during process chromatography, we demonstrate this process can be 'fine tuned' to enhance target antibody selectivity by removing host cell protein impurities and aggregates. When used in conjunction, these technologies represent a highly efficient process to obtain monoclonal antibodies for use as therapeutics.
Presented as part of the Avantor webinar program in June 2020.
Register & watch
Presented by
joshua-sumoski-100.jpg
Joshua Sumoski
Joshua is an Innovation Scientist at Avantor. In his current role, Joshua focuses on downstream purification process development and chromatography product support. He has over 15 years of experience within the biotech/pharma industry in both process development and manufacturing.
Jonathan-Fura-100.jpg
Dr. Jonathan Fura
Previously held: Manager, Research & Development
Jonathan focused on developing technologies that are utilized to advance the development and manufacturing of antibody-based biopharmaceuticals and diagnostics. Jonathan holds a Ph.D. in chemistry with a focus on developing immunomodulation therapeutic strategies.
jungmin-oh-2021-100x100.jpg
Dr. Jungmin Oh
Manager, New Product Development
In her current role, Jungmin leads product and process development projects with multiple biopharmaceutical industry partners, including customized product development for cell and gene therapy customers. She holds a MS and Ph.D. in chemical engineering, specializing in the optimization of a continuous chromatography system.