Ask Avantor
Expert help at your fingertips
Avantor® provides expert support on a wide range of technologies, operational questions and ways in which we empower customer successes in biopharma, advanced technologies, biomaterials, laboratory services and research and diagnostics.
Conversion tables
Key information for the working scientist
As a trusted partner with a passion for science, Avantor® offers a broad array of useful conversion tables and formulas to help your research and production to set science in motion.
Particle size 1
| Sieve "Mesh" | Sieve Opening | |
|---|---|---|
| Inches | Millimeters | |
| 1 | 1.00 | 25.4 |
| 7/8 | 0.875 | 22.6 |
| 3/4 | 0.750 | 19.0 |
| 5/8 | 0.625 | 16.0 |
| .530 | 0.530 | 13.5 |
| 1/2 | 0.500 | 12.7 |
| 7/16 | 0.438 | 11.2 |
| 3/8 | 0.375 | 9.51 |
| 5/16 | 0.312 | 8.00 |
| .265 | 0.265 | 6.73 |
| 1/4 | 0.250 | 6.35 |
| 3 1/2 | 0.223 | 5.66 |
| 4 | 0.187 | 4.76 |
| 5 | 0.157 | 4.00 |
| 6 | 0.132 | 3.36 |
| 7 | 0.111 | 2.83 |
| 8 | 0.0937 | 2.38 |
| 10 | 0.0787 | 2.00 |
| 12 | 0.0661 | 1.68 |
| 14 | 0.0555 | 1.41 |
| 16 | 0.0469 | 1.19 |
| 18 | 0.0394 | 1.00 |
| 20 | 0.0331 | 0.841 |
| 25 | 0.0278 | 0.707 |
| 30 | 0.0234 | 0.595 |
| 35 | 0.0197 | 0.500 |
| 40 | 0.0165 | 0.420 |
| 45 | 0.0139 | 0.354 |
| 50 | 0.0117 | 0.297 |
| 60 | 0.0098 | 0.250 |
| 70 | 0.0083 | 0.210 |
| 80 | 0.0070 | 0.177 |
| 100 | 0.0059 | 0.149 |
| 120 | 0.0049 | 0.125 |
| 140 | 0.0041 | 0.105 |
| 170 | 0.0035 | 0.088 |
| 200 | 0.0029 | 0.074 |
| 230 | 0.0025 | 0.063 |
| 270 | 0.0021 | 0.053 |
| 325 | 0.0017 | 0.044 |
| 400 | 0.0015 | 0.037 |
1. Large sieve opening (1 to 1/4 inch) have been designated a sieve "mesh" size that corresponds to the size of the opening in inches.
Sieve "mesh" sizes of 31/2 and higher are designated by the number of openings per linear inch in the sieve.
Note: To convert millimeters to microns, move decimal point three places to the right. Example: 0.125 millimeters converts to 125 microns.
Acid / Base Concentrations
Composition of concentrated reagent-grade acids, ammonium hydroxide, and sodium and potassium hydroxide solutions (with dilution directions to 1M and 1N solutions)
| Chemical | Formula Weight of Reagenta | Approximate Density | Approximate Strength of Concd. Reagentb | Assay Limits % w/w | Molarity of Concd. Reagent | Milliliters of Concd. Reagent Necessary to Prepare 1 Liter of 1 Molar Solutionc | Normality of Concd. Reagent | Milliliters of Concd. Reagent Necessary to Prepare 1 Liter of 1 Normal Solutionc |
|---|---|---|---|---|---|---|---|---|
| Acetic Acid (CH3COOH) | 60.052 | 1.05 | 99.8 | 99.7–99.9 | 17.4 | 57.5 | 17.4 | 57.5 |
| Formic Acid (HCOOH) | 46.026 | 1.13 | 90 | 88.0–92.0 | 23.6 | 42.5 | 23.6 | 42.5 |
| Hydrochloric Acid (HCI) | 36.461 | 1.18 | 37.2 | 36.5–38.0 | 12.1 | 82.5 | 12.1 | 82.5 |
| Hydrofluoric Acid (HF) | 20.006 | 1.19 | 49 | 48.0–51.0 | 28.9 | 34.5 | 28.9 | 34.5 |
| Nitric Acid (HNO3) | 63.013 | 1.41 | 69.6 | 69.0–70.0 | 15.6 | 64 | 15.6 | 63 |
| Perchloric Acid (HCIO4) | 100.458 | 1.67 | 70.5 | 70.0–72.0 | 11.7 | 85.5 | 11.7 | 85.5 |
| Perchloric Acid (HCIO4) | 100.458 | 1.67 | 61.3 | 60.0–62.0 | 9.5 | 105.5 | 9.5 | 105.5 |
| Phosphoric Acid (H3PO4) | 97.995 | 1.71 | 85.5 | 85.0–87.0 | 14.8 | 67.5 | 44.4 | 22.5 |
| Sulfuric Acid (H2SO4) | 98.073 | 1.84 | 96 | 95.0–98.0 | 18 | 55.5 | 36 | 28 |
| Ammonium Hydroxide (NH4OH) | 35.046 | 0.90 | 56.6d | — | 14.5 | 69 | 14.5 | 69 |
| Sodium Hydroxide (NaOH) | 39.997 | 1.53 | 50.5 | 50.0–52.0 | 19.4 | 51.5 | 19.4 | 51.5 |
| Potassium Hydroxide (KOH) | 56.105 | 1.45 | 45 | 45.0–46.0 | 11.7 | 85.5 | 11.7 | 85.5 |
a. Based on atomic weight table (32C = 12).
b. Representative value, w/w %.
c. Rounded to nearest 0.5 ml.
d. Equivalent to 28.0 % w/w NH3.