Orders LinkContactUsLinkComponent
A scientist in full protective gear adjusting tubing connectors on a bioprocess container
Reliable Bioprocess Containers for Storage and Transport
Protect the integrity of your process fluids and biological materials with our bioprocess containers and 3D single-use bags, engineered to ensure safe storage and transport. Choose between an expansive portfolio of plastic and stainless-steel solutions supporting single-use bag assemblies in volumes from 50L to 2500L.
Request a Consultation
Bioprocess Containers for Storage and Transport | Avantor
Comprehensive Suite of Container Solutions
Image of an Avantor stainless steel container
Stainless Steel Containers
Explore durable, stainless-steel containers, ideal for GMP environments and cleanroom transfer. Supporting working volumes of 50L–1500L, these containers are easily configured with load cells, double-jacketed for temperature control, and have bag inflation options.
Image of a transport trolley carrying an Avantor plastic bin
Move heavy loads safely and efficiently with manual and motor-driven dollies and trolleys. Options include load cells and controlling software for advanced operational processing and quality control.
Explore Trolley Options
Image of an Avantor 3D single-use bioprocess bag
We offer standard 3D cubic bags in volumes from 50L to 1500L, and custom bags up to 2500L. Our bioprocess bags have multiple port options constructed with a multi-layer film providing product integrity and robust durability for storage and transport.
View 3D Bioprocessing Bags
Engineered-to-Order Container Solutions 
Engineered-to-order designs supporting volumes up to 2500L, with options for unique geometries and mobility requirements. In addition to standard configurable options, features like leak-proof design and UV shielding are available.
A scientist in full protective gear pushing a trolley carrying an Avantor plastic bin
Protect Quality from Storage to Delivery
Proper storage and transportation of process liquids and biological materials are essential to maintaining quality, safety, and regulatory compliance. Bioprocess containers must perform reliably under pressure, temperature variation, and movement, without compromising material integrity. Avantor provides solutions designed to meet these demands and support consistent performance across your bioprocesses.
Proven Performance Backed by Global Expertise
Icon of a cleanroom
Validated for Critical Use
Our bioprocess containers are designed and tested to meet the demands of regulated environments, including ISTA and UN compatibility standards.
Icon of a globe with a package and arrow representing global support
Consistent Supply Worldwide
With global manufacturing and logistics support, Avantor helps you maintain availability and continuity from pilot scale to commercial production.
Icon of a person wearing a hard hat and a piece of paper with test tubes
Engineered for Your Processes
Standard and custom designs are available in stainless steel or polymer, with configurable features that match your process and compliance needs.
Featured Products
Image of several Avantor single-use mixers
Reduce contamination risk and improve efficiency with scalable, single-use mixing systems, configurable to your production requirements.
View Mixing Systems
Image of an Avantor custom bioprocess system
Design integrated, single-use, or hybrid systems tailored to your processes. Avantor’s custom assemblies and components support sterile fluid transfer, scale-up, and cGMP compliance.
Explore our Custom Bioprocess Systems
A scientist in full protective gear in a buffer management facility
Avantor’s hydration platform simplifies solution preparation with critical reagent-grade buffers, concentrates, and media in scalable formats. 
Browse Hydrated Buffers
Talk To An Expert
Whether you need standard bioprocess containers or customized designs, our team can help guide selection, validation, and implementation. Connect with an expert to find the right fit for your process and compliance needs.
Talk To An Expert
Featured Resources
Icon of several reports
Discover our full range of single-use systems, from fluid handling and mixing to buffer management; all designed for scalable, compliant production.
Download Brochure
Icon of a piece of paper with a magnifying glass
Review Avantor’s validation guide to understand compliance, testing, and manufacturing standards for single-use bioprocess bags.
View the Guide
Icon of a book
Access our complete library of brochures, datasheets, and technical papers for bioprocess containers, assemblies, and other single-use solutions.
Explore Library