Orders LinkContactUsLinkComponent
method 533 heromain page.jpg
EPA Method 533 for Drinking water
Determination of selected PFAS in Drinking water by Isotope dilution anion exchange solid phase extraction and Liquid Chromatography/tandem mass spectrometry (LC-MS/MS.
EPA Method 533 | Avantor
1stproductsamplepreparation.jpg
PFAS friendy vials
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
Caps for PFAS analysis
2stproductsamplepreparation.jpg
SPE for PFAS analysis
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.
2stproductsamplepreparation.jpg
LC Columns
2stproductsamplepreparation.jpg
GC Columns