Order Entry
France
ContactUsLinkComponent
MethoCult™ H4434 Classic Medium
MethoCult™ H4434 Classic Medium
Référence STMC04444
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
undefined
MethoCult™ H4434 Classic Medium
Référence STMC04444
Fournisseur:  STEMCELL Technologies
Numéro CAS:  
Spécifications
  • Cdt:
    72 mL
  • Désignation:
    MethoCult™ H4434 Classic
Spécifications
À propos de cet article
Pre-aliquoted methylcellulose-based medium with recombinant cytokines for human cells. 24×3 ml vials.
MethoCult™ H4434 Classic (MethoCult™ GF H4434) is a complete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. MethoCult™ H4434 Classic is formulated to support the optimal growth of erythroid progenitor cells (BFU-E and CFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G and CFU-M), and multipotential granulocyte, erythroid, macrophage and megakaryocyte progenitor cells (CFU-GEMM).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').