1250 UL PIPETTE TIP UNIV FIT 1 * 10 items
Catalog # POCA1PT1250FOU
Supplier: Polycarbin
New Product
Frequently Bought Together
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: VWR Collection
Water standards, CRM, Karl Fischer titration, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
VWR®, Speciality pH Electrodes
Supplier: VWR Collection
Saucepans with handle
Supplier: MORGAN ADVANCED MATERIALS HALD
Benzene, Supelco®
Supplier: Merck
Nitric acid 0.2 mol/l (0.2 N)
Supplier: VWR Collection