Order Entry
France
ContactUsLinkComponent
VWR Collection Aluminium Weigh Boat, Certified Oil Free, 25ml, 52 x 13mm crinkle wall, pk100 1 * 100 items
Catalog # HEAT120931FRV
Supplier:  HEATHROW
New Product
VWR Collection Aluminium Weigh Boat, Certified Oil Free, 25ml, 52 x 13mm crinkle wall, pk100 1 * 100 items
Catalog # HEAT120931FRV
Supplier:  HEATHROW