Order Entry
France
ContactUsLinkComponent
Primary standards Potassium Hydrogen Phthalate CRM, ISO 17034 and ISO 17025,,,, Conservation 36 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 50g bottle 1 * 50 g
Catalog # ACSDBP647.50G
Supplier:  CPACHEM
New Product
Primary standards Potassium Hydrogen Phthalate CRM, ISO 17034 and ISO 17025,,,, Conservation 36 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,, 50g bottle 1 * 50 g
Catalog # ACSDBP647.50G
Supplier:  CPACHEM