Order Entry
France
ContactUsLinkComponent
A heterodimer of Integrin alpha V (Phe31-Val992) with a C-terminal 6xHis tag and Integrin beta 1 (Gln21-Asp728) with a C-terminal hu man Fc tag. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA317942-100
Fournisseur:  ANTIBODIES.COM
New Product
A heterodimer of Integrin alpha V (Phe31-Val992) with a C-terminal 6xHis tag and Integrin beta 1 (Gln21-Asp728) with a C-terminal hu man Fc tag. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA317942-100
Fournisseur:  ANTIBODIES.COM