Order Entry
France
ContactUsLinkComponent
Dipyrone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Dipyrone in raw comm ingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327200-96
Supplier:  ANTIBODIES.COM
New Product
Dipyrone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Dipyrone in raw comm ingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327200-96
Supplier:  ANTIBODIES.COM