Order Entry
France
ContactUsLinkComponent
Beta Lactam + Cephalexin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Beta Lactam + Cephalexin in milk. 1 * 96 Tests
Catalog # ANTIA327181-96
Supplier:  ANTIBODIES.COM
New Product
Beta Lactam + Cephalexin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Beta Lactam + Cephalexin in milk. 1 * 96 Tests
Catalog # ANTIA327181-96
Supplier:  ANTIBODIES.COM