Order Entry
France
ContactUsLinkComponent
Sulfonamide + Fluoroquinolones + Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Sulfonamide + Fluoroquinolones + Tetracycline in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327254-96
Supplier:  ANTIBODIES.COM
New Product
Sulfonamide + Fluoroquinolones + Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Sulfonamide + Fluoroquinolones + Tetracycline in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327254-96
Supplier:  ANTIBODIES.COM