Order Entry
France
ContactUsLinkComponent
Dexamethasone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Dexamethasone i n raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327199-96
Supplier:  ANTIBODIES.COM
New Product
Dexamethasone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Dexamethasone i n raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327199-96
Supplier:  ANTIBODIES.COM