SODIUM SULFIDE NONAHYDRATE 98% 1 * 25 g
Catalog # APOSIN3316-25G
Supplier: Apollo Scientific
New Product
SODIUM SULFIDE NONAHYDRATE 98% 1 * 25 g
Catalog # APOSIN3316-25G
Supplier: Apollo Scientific
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Hydrochloric acid 2 mol/l (2 N), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Nitrile Extra Light blue Nitrile Gloves
Supplier: VWR Collection