HighFlow Solution System: - XL funnel with lid, OD 200 mm, Material: PE-HD-EL, including coarse sieve, Thread: male thread M34x2 - Main pipe, OD 24 mm, ID 20 mm, length approx. 800 mm (Material: stainless steel 1.4571 (V4A), Thread: female thread M34 1 * 1 items
Catalog # SCTV106623
Supplier: S.C.A.T.
HighFlow Solution System: - XL funnel with lid, OD 200 mm, Material: PE-HD-EL, including coarse sieve, Thread: male thread M34x2 - Main pipe, OD 24 mm, ID 20 mm, length approx. 800 mm (Material: stainless steel 1.4571 (V4A), Thread: female thread M34 1 * 1 items
Catalog # SCTV106623
Supplier: S.C.A.T.