Order Entry
France
ContactUsLinkComponent
Description,, Detection instrument: Microplate reader(345 nm - 360 nm, optimum wavelength: 355 nm) Assay time: 40 min Standard Curve Range: 16 - 641 U/L Sensitivity: 16 U/L Inter-Assay,,%CV < 6 Intra-Assay,,%CV < 3.3 Spike Recovery,, Average recovery 1 * 96 Assays
Catalog # NOVUNBP3-24520
Supplier:  Novus Biologicals
New Product
Description,, Detection instrument: Microplate reader(345 nm - 360 nm, optimum wavelength: 355 nm) Assay time: 40 min Standard Curve Range: 16 - 641 U/L Sensitivity: 16 U/L Inter-Assay,,%CV < 6 Intra-Assay,,%CV < 3.3 Spike Recovery,, Average recovery 1 * 96 Assays
Catalog # NOVUNBP3-24520
Supplier:  Novus Biologicals