Order Entry
France
ContactUsLinkComponent
High Precision Bench Scale SCK: PBK9890R2D3M20S200 Commodity Code: 8423828900 Country of Origin: DE Base Model: PBK989 Application: Hazardous area Cat 3GD/Div2 Capacity: 3 kg Platform size: A, 240x300mm IP-Protection: IP66/68 Resolution: 600.000d/750 1 * 1 items
Catalog # METB30214791
Supplier:  Mettler - Toledo
New Product
High Precision Bench Scale SCK: PBK9890R2D3M20S200 Commodity Code: 8423828900 Country of Origin: DE Base Model: PBK989 Application: Hazardous area Cat 3GD/Div2 Capacity: 3 kg Platform size: A, 240x300mm IP-Protection: IP66/68 Resolution: 600.000d/750 1 * 1 items
Catalog # METB30214791
Supplier:  Mettler - Toledo