Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EL. AG/AGCL DJ RN SC 1 * 1 Pce
Catalog # MSPPOF60726100
Supplier:
MARKET SOURCE PART PROCESS
Expand image
EL. AG/AGCL DJ RN SC 1 * 1 Pce
Catalog # MSPPOF60726100
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥99%, KLINIPATH for histology (denatured with 5% methanol)
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals