ALPHATEC 87-190 GLOVES, GLOVE SIZE: 7 1 * 12 PAIRE
Catalog # MSPPOFA132261
Supplier: MARKET SOURCE PART PROCESS
ALPHATEC 87-190 GLOVES, GLOVE SIZE: 7 1 * 12 PAIRE
Catalog # MSPPOFA132261
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Formaldehyde solution Emprove Evolve 1 * 1 L
Supplier: MERCK PRODUCTION CHEMICALS
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF