ZETA POTENTIAL TRANSFER STANDARD 1 BOX 1 * 10 ST
Catalog # MSPPZTS1240
Supplier: MARKET SOURCE PART PROCESS
ZETA POTENTIAL TRANSFER STANDARD 1 BOX 1 * 10 ST
Catalog # MSPPZTS1240
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Bottles, heavy duty, wide neck, with screw cap, for demanding applications, Nalgene®
Supplier: Thermo Fisher Scientific
Bottle top vacuum filtration systems, VacuCap®, Supor® membrane
Supplier: Cytiva (Formerly Pall Lab)
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Masterflex® L/S® Precision Pump Tubing, PharmaPure®, Avantor®
Supplier: Avantor Fluid Handling
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Round cuvettes
Supplier: Hach
Acetic acid 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
Supplier: Honeywell Chemicals