Qualitix® Sterilisation Pouches
Product Details & Documents
Fitting any small to mid-size laboratory, medical, dental or veterinary device, the Qualitix® single use pouches provide easy and effective wrapping of items requiring sterilization.
- Robust durable material
- Quick content identification
- Large self-sealing strip, easy to close
- Chevron top seal for easy opening
- Suitable for steam and EO gas sterilization
- Two sterilization indicators
- Five different sizes
Made with robust medical grade paper, the pouches tightly close with self-sealing strip to maintain sterility. Content remains always visible through PET/CPP transparent
laminated film.
Two chemical indicators included to confirm proper sterilisation process. Blue indicator turns dark grey after autoclaving, red indicator turns to yellow after ethylene oxide (EO) gas sterilisation.
Blue stained laminated film enables easy control of film/pouch integrity. Do not use a damaged pouch.
Certifications: Conformity to ISO 17665-06, ISO 11135, CE.
Documents
Specifications
- Description:Pouch, self-sealing
- L×W:300×490 mm
- Seq:05