12 COMPONENTS: 10MG/L EACH IN METHANOL 1 * 5 Ampoul
Catalog # CMPLRD0619241.5
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
12 COMPONENTS: 10MG/L EACH IN METHANOL 1 * 5 Ampoul
Catalog # CMPLRD0619241.5
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Sodium chloride 0.9% in aqueous solution, OTEC, sterile
Supplier: AGUETTANT
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
ACCUSPIN™ Centrifuge Tubes, Sterile, Polypropylene
Supplier: SIGMA ALDRICH MICROSCOPY
Hydrochloric acid 2 mol/l (2 N)
Supplier: PROLABO CMP
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Supplier: PROLABO CMP
Ammonium formate, Supelco®
Supplier: Merck