Order Entry
France
Orders LinkContactUsLinkComponent
Selective Supplements

Selective Supplements

Bos antibiotic supplement for the selective isolation of Bordetella pertussis, 1
Catalog # HIMEFD004-5X5VL
Supplier:   HIMEDIA
Selective Supplements
Catalog # HIMEFD004-5X5VL
Supplier:  HIMEDIA
CAS Number:  
Selective Supplements

Bos antibiotic supplement for the selective isolation of Bordetella pertussis, 1

CAS Number:  
Price
152,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

Antibiotic supplement recommended for the selective isolation of various microorganisms.

  • Store at 2...8 °C
Specifications

Specifications

  • Pk:
    1
  • Description:
    Bos antibiotic supplement for the selective isolation of <i>Bordetella pertussis</i>

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.