Order Entry
France
Orders LinkContactUsLinkComponent
Objectives
Objectives
Apochromat objective, 0,1 mm
Catalog # OLYMN5702700
Supplier:   OLYMPUS OPTICAL
Objectives
Catalog # OLYMN5702700
Supplier:  OLYMPUS OPTICAL
Objectives
Objectives
Apochromat objective, 0,1 mm
Price
Inquire For Price
Log in for availability
1 * 1 Pce
Product Details & Documents
Specifications
  • Magnification:
    100X
  • Description:
    Apochromat objective
  • Seq:
    08
  • Working distance:
    0,1 mm