Order Entry
France
ContactUsLinkComponent
FILTER INTERFERENCE 10*7.5MM 620NM 1 * 1 items
Catalog # TCAT13714409
Supplier:  Tecan
New Product
FILTER INTERFERENCE 10*7.5MM 620NM 1 * 1 items
Catalog # TCAT13714409
Supplier:  Tecan