Order Entry
France
ContactUsLinkComponent
Minivette® POCT Lithium heparin LH, nominal volume: 200 µl, preparation: Heparin, orange, colour code EU, 150 piece(s)/bag, 150 piece(s)/case 1 * 1.500 items
Catalog # SARS17.2112.200
Supplier:  SARSTEDT
Minivette® POCT Lithium heparin LH, nominal volume: 200 µl, preparation: Heparin, orange, colour code EU, 150 piece(s)/bag, 150 piece(s)/case 1 * 1.500 items
Catalog # SARS17.2112.200
Supplier:  SARSTEDT
Frequently Bought Together