Ac-DEVD-CHO (Trifluoroacetate Salt) 1 * 5 mg
Catalog # STMC100-0537
Supplier: STEMCELL Technologies
Frequently Bought Together
Methyl ethyl ketone ≥99.7%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane, anhydrous (max. 0.003% H₂O)
Supplier: VWR Chemicals
Anisole, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, anhydrous ≥99.8% unstabilised
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% ACS
Supplier: Thermo Fisher Scientific
Butanone (Methylethylketone) (Reag. USP, Ph. Eur.) for analysis, ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
Dimethyl sulphoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND