MOUSSE HYDROALCOOLIQUE InstantFOAM COMPLETE 1 * 6 Pce
EVONIAF250ML
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
: MERCK PRODUCTION CHEMICALS
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
Petroleum spirit, 30…50 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2,2,4-trimethylpentane ar, ar®, general lab reagent 1 * 25 L
: MACRON AVANTOR BRAND
Chloroform ≥99.5% stabilised
: Honeywell Chemicals