SONIDEGIB DIPHOSPHATE 1 * 500 mg
Catalog # ENZOENZCHM2370500
Supplier: Enzo Life Sciences
Frequently Bought Together
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Chromium(III) chloride hexahydrate 98-101%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Calcium chloride dihydrate 97.0-103.0% Ph. Eur.
Supplier: VWR Chemicals
Sodium hexanitritocobaltate(III), Supelco®
Supplier: Merck
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium hydrogen phthalate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Lead(II) acetate trihydrate 99.5-103.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Strontium chloride hexahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Magnesium ≥99%, turnings
Supplier: Thermo Fisher Scientific
Manganese(II) sulphate monohydrate ≥99%
Supplier: Thermo Fisher Scientific