CD79A RECOMBINANT MONOCLONAL ANTIBODY (IGA/1790R) 1 * 100 µG
ENZOENZABS3340100
Frequently Bought Together
VWR®, Breatheable Rayon Film Seals for Biological Cultures
: VWR Collection
Multiwell cell culture plates, Falcon®
: Corning
VWR®, High Performance Blender Bags
: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
: VWR Collection
Sampling scoops, SteriPlast®
: BURKLE
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
Serological pipettes, Stripette™
: Corning
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
External thread cryogenic vials
: Corning
VWR®, Petri Dish, PS
: VWR Collection
J.T.Baker®, Plates, Square Well
: Avantor
Accessories and consumables for KingFisher™ Flex
: Thermo Fisher Scientific
Blotting transfer papers
: Ahlstrom-Munksjö
PCR Thermo-Tube and cap strips, 0.2 ml, ABgene®
: Thermo Fisher Scientific