Order Entry
France
Orders LinkContactUsLinkComponent
Fmoc-Sta-OH ≥95%

Fmoc-Sta-OH ≥95%

Plastic bottle, 10 g
Catalog # AATB5107
Irritant
Supplier:   AAT Bioquest
Fmoc-Sta-OH ≥95%
Catalog # AATB5107
Supplier:  AAT Bioquest
CAS Number:  158257-40-0
Fmoc-Sta-OH ≥95%
Fmoc-Sta-OH ≥95%

Plastic bottle, 10 g

CAS Number:  158257-40-0
Price
4 660,00 €
Log in for availability
1 * 10 g
Product Details & Documents

(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.

Synonyms:  (3S,4S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxy-6-methylheptanoic acid
  • CAS Number:158257-40-0
  • Formula:C₂₃H₂₇NO₅
  • MDL Number:MFCD00065675
  • Molecular Weight:397.48 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.