(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic acid certified reference material, TraceCERT(R) 1 * 25 mg
PHR9105-25MG
: Merck
(2R)-2-[(2,2-Dimethylpropanoyl)amino]-2-phenylacetic acid certified reference material, TraceCERT(R) 1 * 25 mg
PHR9105-25MG
: Merck
Frequently Bought Together
VWR®, General purpose viscosity standards
: VWR Chemicals
VWR®, TOC standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
Colour Reference Solutions, Reag.Ph.Eur.
: REAGECON
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Buffer solutions in sachets
: Mettler - Toledo
BUFFER SOLUTION PH4 1 * 1 L
: FF CHEMICALS