Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Fexofenadine methyl ester certified reference material, TraceCERT(R) 1 * 50 mg
Catalog # PHR9103-50MG
Supplier:
Merck
Expand image
Fexofenadine methyl ester certified reference material, TraceCERT(R) 1 * 50 mg
Catalog # PHR9103-50MG
Supplier:
Merck
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Chloride standard solution, 1,000 mg/L Cl- in Water (from NaCl) Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Supplier: Merck
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ