4-(Diethylamino)but-2-ynyl 2-hydroxy-2,2-diphenylacetate certified reference material, TraceCERT(R) 1 * 100 mg
Catalog # PHR9095-100MG
Supplier: Merck
4-(Diethylamino)but-2-ynyl 2-hydroxy-2,2-diphenylacetate certified reference material, TraceCERT(R) 1 * 100 mg
Catalog # PHR9095-100MG
Supplier: Merck
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Calcium standard for ISE calibration, Orion™
Supplier: Thermo Orion
Boron Standard for AAS 1 * 250 mL
Supplier: Merck
(Aminomethyl)phosphonic acid
Supplier: EHRENSTORFER
N-Nitroso-di-n-butylamine certified 1 * 0,1 g
Supplier: EHRENSTORFER
Testosterone, analytical standard, VETRANAL™, Supelco®
Supplier: Merck
Molybdenum standard solution, for ICP, in 5% HNO₃, VHG
Supplier: LGC Standards PROMOCHEM
UBIQUINOL 1 * 1 mL
Supplier: Merck
Dimethyl disulphide
Supplier: EHRENSTORFER
Indium Standard for ICP 1 * 100 mL
Supplier: Merck
4-Nonylphenol-ethoxylate (mono-, di-, tri-) (technical) certified 1 * 0,25 g
Supplier: EHRENSTORFER
Saccharin, Supelco®
Supplier: Merck