N-Nitrosomethylaminobutyric Acid (NMBA) certified reference material, pharmaceutical secondary standard 1 * 1 mL
PHR3610-1ML
: Merck
N-Nitrosomethylaminobutyric Acid (NMBA) certified reference material, pharmaceutical secondary standard 1 * 1 mL
PHR3610-1ML
: Merck
Frequently Bought Together
Triflumizole certified 1 * 0,1 g
: EHRENSTORFER
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution
: Thermo Fisher Scientific
Thiofanox-sulfoxide certified 1 * 0,1 g
: EHRENSTORFER
3,3,5-Trimethylcyclohexyl salicylate certified 1 * 0,25 g
: EHRENSTORFER