0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Acetaldehyde-2,4-DNPH pharmaceutical secondary standard, certified reference material 1 * 100 mg
PHR3523-100MG
:
Merck
Acetaldehyde-2,4-DNPH pharmaceutical secondary standard, certified reference material 1 * 100 mg
PHR3523-100MG
:
Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals