COCONUT OIL, 1000MG, NEAT certified reference material, pharmaceutical secondary standard 1 * 1 g
Catalog # PHR3520-1G
Supplier: Merck
COCONUT OIL, 1000MG, NEAT certified reference material, pharmaceutical secondary standard 1 * 1 g
Catalog # PHR3520-1G
Supplier: Merck
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffer solutions, colour coded
Supplier: Hach
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards
Supplier: HAMILTON BONADUZ
BUFFER PH 7,00 DOSING BOTTLE DURACAL 1 * 500 mL
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
VWR®, Cold Cranking Simulator (CCS) viscosity standards
Supplier: VWR Chemicals
pH Buffer solutions
Supplier: Hach