0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Doxazosin Related Compound A certified reference material, pharmaceutical secondary standard 1 * 50 mg
PHR3313-50MG
:
Merck
Doxazosin Related Compound A certified reference material, pharmaceutical secondary standard 1 * 50 mg
PHR3313-50MG
:
Merck
Frequently Bought Together
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
pH buffer solutions
: Mettler - Toledo
Buffer solutions in sachets
: Mettler - Toledo
Nitrite standard solution, 1000 mg/L NO₂- in Water (from NaNO₂) Certipur®, Supelco®
: Merck
pH buffers, ready to use
: WTW
VWR®, Conductivity standard solutions
: VWR Chemicals