Pindolol pharmaceutical secondary standard, certified reference material 1 * 500 mg
Catalog # PHR3224-500MG
Supplier: Merck
Pindolol pharmaceutical secondary standard, certified reference material 1 * 500 mg
Catalog # PHR3224-500MG
Supplier: Merck
Frequently Bought Together
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Colour Reference Solutions, Reag.Ph.Eur.
Supplier: REAGECON
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Xylenol orange triturate 1% (w/w) in potassium nitrate Reag. Ph. Eur. 1096301
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
CONDUCTIVITY STANDARD 1.3µS/CM 1 * 250 mL
Supplier: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Refractive index standards according to Ph. Eur., Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
PARACETAMOL IMPURITY J 1 * 15 mg
Supplier: EDQM
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo